Triethyl 2-phosphonopropionate, 98%, Thermo Scientific Chemicals
Triethyl 2-phosphonopropionate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethyl 2-phosphonopropionate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 174360100
Price (USD)/ Each
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3699-66-9
IUPAC Nameethyl 2-(diethoxyphosphoryl)propanoate
Molecular FormulaC9H19O5P
InChI KeyBVSRWCMAJISCTD-UHFFFAOYNA-N
SMILESCCOC(=O)C(C)P(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Appearance (Color)Clear colorless
Refractive index1.4310 to 1.4330 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only