Thermo Scientific Chemicals

Triethyl 2-phosphonopropionate, 98%, Thermo Scientific Chemicals

Catalog number: 174360500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Triethyl 2-phosphonopropionate, 98%, Thermo Scientific Chemicals

Catalog number: 174360500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 174360500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3699-66-9
IUPAC Name
ethyl 2-(diethoxyphosphoryl)propanoate
Molecular Formula
C9H19O5P
InChI Key
BVSRWCMAJISCTD-UHFFFAOYNA-N
SMILES
CCOC(=O)C(C)P(=O)(OCC)OCC
Appearance (Form)
Liquid
GC
>=97.5 %
Appearance (Color)
Clear colorless
Refractive index
1.4310 to 1.4330 (20°C, 589 nm)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text