Thermo Scientific Chemicals

Nitrosonium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog number: 174390050
5 g, Plastic bottle, Each
Thermo Scientific Chemicals

Nitrosonium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog number: 174390050
5 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 174390050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14635-75-7
IUPAC Name
dioxoamine; tetrafluoroboranuide
Molecular Formula
BF4HNO2
InChI Key
XYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES
[O-][NH+]=O.F[B-](F)(F)F
Appearance (Form)
Crystalline powder or crystals
Appearance (Color)
White
Assay
>=96.0 % (ex F)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text