Thermo Scientific Chemicals

4-Hydroxy-3-methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 174580010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

4-Hydroxy-3-methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 174580010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 174580010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
498-00-0
IUPAC Name
4-(hydroxymethyl)-2-methoxyphenol
Molecular Formula
C8H10O3
InChI Key
ZENOXNGFMSCLLL-UHFFFAOYSA-N
SMILES
COC1=CC(CO)=CC=C1O
Appearance (Form)
Crystalline powder
Appearance (Color)
Off-white to beige
Infrared spectrum
Conforms
GC
>=98.5 %
Melting point
112°C to 115°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text