4-Hydroxy-3-methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals
4-Hydroxy-3-methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxy-3-methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number 174582500
Price (USD)/ Each
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-(hydroxymethyl)-2-methoxyphenol
Molecular FormulaC8H10O3
InChI KeyZENOXNGFMSCLLL-UHFFFAOYSA-N
SMILESCOC1=CC(CO)=CC=C1O
Molecular Weight (g/mol)154.17
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Off-white to beige
Infrared spectrumConforms
GC>=98.5 %
Melting point112°C to 115°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only