Thermo Scientific Chemicals

Methyltriethoxysilane, 98+%, Thermo Scientific Chemicals

Catalog number: 174622500
250 mL, Glass bottle, Each
Thermo Scientific Chemicals

Methyltriethoxysilane, 98+%, Thermo Scientific Chemicals

Catalog number: 174622500
250 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 174622500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2031-67-6
IUPAC Name
triethoxy(methyl)silane
Molecular Formula
C7H18O3Si
InChI Key
CPUDPFPXCZDNGI-UHFFFAOYSA-N
SMILES
CCO[Si](C)(OCC)OCC
Appearance (Color)
Clear colorless
Refractive index
1.3825 to 1.3845 (20°C, 589 nm)
Appearance (Form)
Liquid
GC
>=98.0 %
Hydrolyzable chloride
=<0.01 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text