Thermo Scientific Chemicals

Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: 174642500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Phenyltriethoxysilane, 98%, Thermo Scientific Chemicals

Catalog number: 174642500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 174642500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
780-69-8
IUPAC Name
triethoxy(phenyl)silane
Molecular Formula
C12H20O3Si
InChI Key
JCVQKRGIASEUKR-UHFFFAOYSA-N
SMILES
CCO[Si](OCC)(OCC)C1=CC=CC=C1
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.4594 to 1.4614 (20°C, 589 nm)
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text