Silicone oil, high temperature
Silicone oil, high temperature
Silicone oil, high temperature
Silicone oil, high temperature
Thermo Scientific Chemicals

Silicone oil, high temperature

CAS: 63148-58-3
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Catalog number 174660025
Price (USD)
939.65
Online Exclusive
1,044.00
Save 104.35 (10%)
Each
In stock
Add to cart
Quantity:
2.5 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
939.65
Online Exclusive
1,044.00
Save 104.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS63148-58-3
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4940 to 1.5040 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless to straw yellow
Appearance (Form)Oily liquid
Viscosity95 to 105 mm²/s (25°C)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection