Thermo Scientific Chemicals

Silicone oil, high temperature, Thermo Scientific Chemicals

Catalog number: 174665000
500 g, Glass Bottle, Each
Thermo Scientific Chemicals

Silicone oil, high temperature, Thermo Scientific Chemicals

Catalog number: 174665000
500 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 174665000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
63148-58-3
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to straw yellow
Appearance (Form)
Oily liquid
Viscosity
95 to 105 mm²/s (25°C)
Refractive index
1.4940 to 1.5040 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text