Thermo Scientific Chemicals

Acetic anhydride-d6, for NMR, 98.5 atom % D, Thermo Scientific Chemicals

Catalog number: 174670050
5 mL, Glass Bottle, Each
Thermo Scientific Chemicals

Acetic anhydride-d6, for NMR, 98.5 atom % D, Thermo Scientific Chemicals

Catalog number: 174670050
5 mL, Glass Bottle, Each
Quantity
Packaging
Catalog number: 174670050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
16649-49-3
IUPAC Name
(2,2,2-²H₃)acetyl (²H₃)acetate
Molecular Formula
C4H6O3
InChI Key
WFDIJRYMOXRFFG-WFGJKAKNSA-N
SMILES
[2H]C([2H])([2H])C(=O)OC(=O)C([2H])([2H])[2H]
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
D-Enrichment
>=98.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text