Methyl sulfoxide-d6, for NMR, +99% atom D, Thermo Scientific Chemicals
Methyl sulfoxide-d6, for NMR, +99% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl sulfoxide-d6, for NMR, +99% atom D, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 174890500
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
207.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
207.00 
Save 31.35 (15%)
Add to cart
Methyl sulfoxide-d6, for NMR, +99% atom D, Thermo Scientific Chemicals
Catalog number174890500
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
207.00 
Save 31.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2206-27-1
IUPAC Name(²H₃)methanesulfinyl(²H₃)methane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-WFGJKAKNSA-N
SMILES[2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
View more
SpecificationsSpecification SheetSpecification Sheet
D-Enrichment>=99.0 %
Appearance (Form)Liquid
Water=<0.1 % (Coulometric)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only