Nunc™ Dishes with UpCell™ Surface
Save 51% on plasticware essentials and stock your lab to culture with confidence. View promotion details >
Nunc™ Dishes with UpCell™ Surface
Thermo Scientific™

Nunc™ Dishes with UpCell™ Surface

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with Thermo Scientific™ Nunc™ Dishes with UpCell™ Surface.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCulture AreaGridded
17490256.7 sq. cmNon-Gridded
17490321.5 sq. cmNon-Gridded
1749048.8 sq. cmNon-Gridded
17490556.7 sq. cmGridded
17490621.5 sq. cmGridded
Catalog number 174902
Price (USD)
639.65
Online Exclusive
682.00
Save 42.35 (6%)
Case of 6
-
Add to cart
Culture Area:
56.7 sq. cm
Gridded:
Non-Gridded
Recurring order eligible. Learn more »
Price (USD)
639.65
Online Exclusive
682.00
Save 42.35 (6%)
Case of 6
Add to cart
Ask our AI about this Product

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of cell sheet with the help of a supportive membrane.

Features:
  • The hydrophicility of the UpCell surface changes with the external temperature, allowing cells to detach
  • No need for dissociation enzymes, ensuring intact cell surface proteins
  • No need for physical scraping, resulting in high cell viability
  • 3 x 3mm grid design in the 60mm and 100mm dishes prevents cells from clumping and enables harvesting of single cell suspension
  • Supportive membranes included in the 35mm dishes allow for harvesting of the cell sheets
  • Shelf Life: 3 years from date of manufacturing

Recommended for:

  • Culture passaging
  • Subsequent single-cell analyses (e.g. flow cytometry)
  • 3D tissue modeling for transplantation
  • For research use only
Warranty: 90 days

 

WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only
Specifications
DescriptionUpCell Dish
Diameter (Metric) Outer100 mm
GriddedNon-Gridded
Height (Metric)17 mm
No. per Case6
No. per Pack1
Surface TreatmentUpCell, temperature responsive
Well ShapeRound, Flat Bottom (F-well)
Culture Area56.7 sq. cm
SterilitySterile
TypeCell Culture Dish
Volume (Metric) Well12.5 mL (Well)
Volume (Metric) Working12.5 mL
Unit SizeCase of 6
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
NU60-2433Certificate of ComplianceDec 10, 2024174903
NU10-2404Certificate of ComplianceDec 06, 2024174902
NU10-2126Certificate of ComplianceDec 06, 2024174902
NU10-2113Certificate of ComplianceDec 06, 2024174902
NU10-2022Certificate of ComplianceDec 06, 2024174902
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

The UpCell Surface is composed of a covalently bound polymer that changes from being slightly hydrophobic to being very hydrophilic when the temperature is reduced to less than 32 degrees C. This results in the binding of water to the surface, swelling of the surface, and finally the release of the cells.

The temperature-responsive polymer is covalently bound to the polystyrene dish or plate and will not detach from the product under normal cell culture procedures and conditions. The products have successfully passed the USP biological reactivity class VI testing.

The “UpCell surface with grid” products have a 3 x 3 mm grid design over the entire culture surface. Cell attachment and growth is restricted to within these small squares, preventing cells from clumping, and enabling easier harvest of the cells as single cells or small cell clusters. Products with “UpCell surface with grid” are suitable for the preparation of cells for flow cytometry analysis but cannot be used for cell-sheet applications. We offer 60 mm and 100 mm dishes with “UpCell surface with grid” whereas the 35 mm dishes contain supportive membrane for harvesting of cell sheets.

You can use the UpCell Surface with many cell types, including fibroblasts, mesenchymal stem cells, myocytes, keratinocytes, macrophages, and different epithelium and tumor cells. If cells do not detach spontaneously from the UpCell surface by lowering of the temperature to below 32 degrees C, the cells can be detached by flushing the surface with medium or by tapping on the side of the dish or plate. Cells that produce a lot of extracellular matrix and attach very strongly to the UpCell surface may be difficult to detach by temperature reduction alone. For these cell types, the simultaneous use of diluted trypsin is recommended.

Coating of the UpCell surface with biomolecules, for example, collagen, fibronectin, poly-lysine and laminin, is possible and should be done if you experience difficulties in attaching your cells to the UpCell surface. The coating procedure should be performed at 37 degrees C.

Share catalog number, name or link

1x1 image pixel for data collection