Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
174927 | Case of 8 |
Significant qualities of the Nunclon Sphera culture surface:
Optimizing spheroid formation
Growing 3D cancer cultures
Promoting formation of embryoid bodies (EB)
Share catalog number, name or link