Mercuric acetate, 98%, Thermo Scientific Chemicals
Mercuric acetate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Mercuric acetate, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 175025000
Price (USD)/ Each
391.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
460.00 
Save 68.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
391.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
460.00 
Save 68.35 (15%)
Add to cart
Mercuric acetate, 98%, Thermo Scientific Chemicals
Catalog number175025000
Price (USD)/ Each
391.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
460.00 
Save 68.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1600-27-7
IUPAC Namemercury(2+) diacetate
Molecular FormulaC4H6HgO4
InChI KeyBRMYZIKAHFEUFJ-UHFFFAOYSA-L
SMILES[Hg++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Complexometric>=97.5 %
Sulfate (SO4)=<100 ppm
Appearance (Color)White to almost white
Melting point179°C to 182°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only