B-27™ Supplement (50X), serum free
B-27™ Supplement (50X), serum free
Gibco™

B-27™ Supplement (50X), serum free

The most cited neuronal cell culture supplement, B-27 Supplement is an optimized serum-free supplement used to support the low- or深入閱讀
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
1750404410 mL
17504001100 mL
產品號碼 17504044
價格 (HKD)
1,125.00
Each
新增至購物車
Quantity:
10 mL
價格 (HKD)
1,125.00
Each
新增至購物車
Ask our AI about this Product
The most cited neuronal cell culture supplement, B-27 Supplement is an optimized serum-free supplement used to support the low- or high-density growth and short- or long-term viability of embryonic, post-natal, and adult hippocampal and other CNS neurons. B-27 Supplement is provided as a 50X liquid and is intended for use with Neurobasal Medium or Neurobasal-A Medium for neuronal cell culture without the need for an astrocyte feeder layer.

B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of B-27 Supplement and Neurobasal media.

For established protocols using B-27 Supplement, access our Gibco Neurobiology Protocol Handbook.

View publications using Gibco neurobiology products at our Gibco CellCite.
For Research Use Only. Not for use in diagnostic procedures.
規格
Cell TypeNeural Cells
Concentration50 X
Culture TypeMammalian Cell Culture
Product LineB-27™
Quantity10 mL
Serum LevelSerum-Free
Shelf Life12 Months
Shipping ConditionDry Ice
SourceSerum-Free
Tested ForEndotoxin, Performance, Sterility, Mycoplasma
ClassificationSerum-free
FormLiquid
Product TypeCell Culture Supplement
SterilitySterile
pH6 to 8
Unit SizeEach
內容物與存放
Store in freezer (-5 to -20°C) and protect from light.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

文件與下載

證書

安全資料表

分享目錄編號、產品名稱或連結。

1x1 image pixel for data collection