Thermo Scientific Chemicals

Trifluoromethanesulfonic anhydride, 98+%, Thermo Scientific Chemicals

Catalog number: 175060500
50 mL, Glass bottle, Each
Thermo Scientific Chemicals

Trifluoromethanesulfonic anhydride, 98+%, Thermo Scientific Chemicals

Catalog number: 175060500
50 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 175060500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
358-23-6
Appearance (Form)
Liquid
Refractive index
1.3200 to 1.3230 (20°C, 589 nm)
Impurity
=<1.0 % CF3SO3H
Titration with CHA
98.0 to 101.0 %
Impurity
=<300 ppm Free fluoride (as F)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent, Coupling reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text