Thermo Scientific Chemicals

Polymethylhydrosiloxane, Thermo Scientific Chemicals

Catalog number: 175091000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Polymethylhydrosiloxane, Thermo Scientific Chemicals

Catalog number: 175091000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 175091000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9004-73-3
Molecular Formula
(CH4OSi)n
SMILES
C[SiH](-*)O-*
MDL Number
MFCD00084478
Infrared spectrum
Conforms
Appearance (Form)
Viscous liquid
Refractive index
1.3970 to 1.4010 (20°C, 589 nm)
Appearance (Color)
Clear colorless
Viscosity
15 to 40 mPa.s (at 20°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text