L(+)-Asparagine monohydrate, 99%
L(+)-Asparagine monohydrate, 99%
Thermo Scientific Chemicals

L(+)-Asparagine monohydrate, 99%

L(+)-Asparagine monohydrate, 99%, C4H10N2O4, CAS Number-5794-13-8, asparagine monohydrate, l +-asparagine monohydrate, asparagine, asparagine nf, l-asparagine hydrate, h-asn-oh.h2o, unii-2pd79vf521, l-asparagine monohydrate, l-asparagine, monohydrate, s-2,4-diamino-4-oxobutanoic acid hydrate, 250g
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
View all
Glass Bottle
Catalog number 175271000
Price (USD)/ Each
83.65
Online exclusive
92.80 
Save 9.15 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
83.65
Online exclusive
92.80 
Save 9.15 (10%)
Add to cart
L(+)-Asparagine monohydrate, 99%
Catalog number175271000
Price (USD)/ Each
83.65
Online exclusive
92.80 
Save 9.15 (10%)
-
Add to cart
Chemical Identifiers
CAS5794-13-8
IUPAC Name(2S)-2-amino-3-carbamoylpropanoic acid hydrate
Molecular FormulaC4H10N2O4
InChI KeyRBMGJIZCEWRQES-DKWTVANSSA-N
SMILESO.N[C@@H](CC(N)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Other amino acids=<1 % (TLC)
Loss on drying11.5 % to 12.5 % (130°C, 3 hrs)
Arsenic (As)=<1 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only