Thermo Scientific Chemicals

Deuterium chloride, for NMR, 20 wt. % solution in D2O, 100.0 atom % D, Thermo Scientific Chemicals

Catalog number: 175340050
5 g, Glass Bottle, Each
Thermo Scientific Chemicals

Deuterium chloride, for NMR, 20 wt. % solution in D2O, 100.0 atom % D, Thermo Scientific Chemicals

Catalog number: 175340050
5 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 175340050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7698-05-7
IUPAC Name
proton chloride
Molecular Formula
ClH
InChI Key
VEXZGXHMUGYJMC-FTGQXOHASA-N
SMILES
[1H+].[Cl-]
D-Enrichment
>=99.95 %
Appearance (Form)
Solution
Appearance (Color)
Clear colorless
Concentration
20 to 22 wt%

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text