Thermo Scientific Chemicals

Sodium deuteroxide, for NMR, 40 wt.% solution in D2O, 99+% atom D, Thermo Scientific Chemicals

Catalog number: 175470500
50 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Sodium deuteroxide, for NMR, 40 wt.% solution in D2O, 99+% atom D, Thermo Scientific Chemicals

Catalog number: 175470500
50 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 175470500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
14014-06-3
IUPAC Name
sodium hydroxide
Molecular Formula
HNaO
InChI Key
HEMHJVSKTPXQMS-UHFFFAOYSA-M
SMILES
[OH-].[Na+]
Appearance (Color)
Clear colorless
Concentration
40 to 42 % w/w
D-Enrichment
>=99.0 %
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text