Thermo Scientific Chemicals

2-Iodoethanol, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 175500500
50 mL, Each
Thermo Scientific Chemicals

2-Iodoethanol, 99%, stabilized, Thermo Scientific Chemicals

Catalog number: 175500500
50 mL, Each
Quantity
Catalog number: 175500500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
624-76-0
IUPAC Name
2-iodoethan-1-ol
Molecular Formula
C2H5IO
InChI Key
QSECPQCFCWVBKM-UHFFFAOYSA-N
SMILES
OCCI
Appearance (Form)
Liquid
GC
>=98.5 %
Refractive index
1.5690 to 1.5720 (20°C, 589 nm)
Appearance (Color)
Clear colorless to yellow-brown
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text