Thermo Scientific Chemicals

2,4-Dihydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: 175590050
5 g, Each
Thermo Scientific Chemicals

2,4-Dihydroxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: 175590050
5 g, Each
Quantity
Catalog number: 175590050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
626-03-9
IUPAC Name
2-hydroxy-1,4-dihydropyridin-4-one
Molecular Formula
C5H5NO2
InChI Key
ZEZJPIDPVXJEME-UHFFFAOYSA-N
SMILES
OC1=CC(=O)C=CN1
Appearance (Color)
White to yellow to light beige
Infrared spectrum
Conforms
Appearance (Form)
Crystalline powder or crystals
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text