3-Quinolinecarboxylic acid, 98%, Thermo Scientific Chemicals
3-Quinolinecarboxylic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Quinolinecarboxylic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 175700010
Price (USD)/ Each
45.60
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
45.60
Add to cart
3-Quinolinecarboxylic acid, 98%, Thermo Scientific Chemicals
Catalog number175700010
Price (USD)/ Each
45.60
-
Add to cart
Chemical Identifiers
CAS6480-68-8
IUPAC Namequinoline-3-carboxylic acid
Molecular FormulaC10H7NO2
InChI KeyDJXNJVFEFSWHLY-UHFFFAOYSA-N
SMILESOC(=O)C1=CN=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Appearance (Form)Powder
Melting point277°C to 283°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only