Thermo Scientific Chemicals

3-Quinolinecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 175700250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Quinolinecarboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: 175700250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 175700250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6480-68-8
IUPAC Name
quinoline-3-carboxylic acid
Molecular Formula
C10H7NO2
InChI Key
DJXNJVFEFSWHLY-UHFFFAOYSA-N
SMILES
OC(=O)C1=CN=C2C=CC=CC2=C1
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
Titration with HClO4
>=97.5 %
Appearance (Form)
Powder
Melting point
277°C to 283°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text