Thermo Scientific Chemicals

Pyruvic aldehyde, 35-45 wt% solution in water, Thermo Scientific Chemicals

Catalog number: 175790025
2.5 kg, Glass bottle, Each
Thermo Scientific Chemicals

Pyruvic aldehyde, 35-45 wt% solution in water, Thermo Scientific Chemicals

Catalog number: 175790025
2.5 kg, Glass bottle, Each
Quantity
Packaging
Catalog number: 175790025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-98-8
IUPAC Name
2-oxopropanal
Molecular Formula
C3H4O2
InChI Key
AIJULSRZWUXGPQ-UHFFFAOYSA-N
SMILES
CC(=O)C=O
Appearance (Form)
Solution
Refractive index
1.3910 to 1.4200 (20°C, 589 nm)
Appearance (Color)
Clear yellow to brown
Titration with NaOH
30 to 45 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text