1,2,3,4-Tetrahydro-1-naphthylamine, 97%, Thermo Scientific Chemicals
1,2,3,4-Tetrahydro-1-naphthylamine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,3,4-Tetrahydro-1-naphthylamine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 175810250
Price (USD)/ Each
174.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
174.00
Add to cart
1,2,3,4-Tetrahydro-1-naphthylamine, 97%, Thermo Scientific Chemicals
Catalog number175810250
Price (USD)/ Each
174.00
-
Add to cart
Chemical Identifiers
IUPAC Name1,2,3,4-tetrahydronaphthalen-1-aminium chloride
Molecular FormulaC10H14ClN
InChI KeyDETWFIUAXSWCIK-UHFFFAOYNA-N
SMILES[Cl-].[NH3+]C1CCCC2=CC=CC=C12
Molecular Weight (g/mol)183.68
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5630 to 1.5650 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow-brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only