4-Acetylbenzoic acid, 98%, Thermo Scientific Chemicals
4-Acetylbenzoic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Acetylbenzoic acid, 98%, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
10 g
Packaging:
Glass bottle
Catalog number 175860100
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
206.00 
Save 30.35 (15%)
-
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
206.00 
Save 30.35 (15%)
Add to cart
4-Acetylbenzoic acid, 98%, Thermo Scientific Chemicals
Catalog number175860100
Price (USD)/ Each
175.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
206.00 
Save 30.35 (15%)
-
Add to cart
Chemical Identifiers
CAS586-89-0
IUPAC Name4-acetylbenzoic acid
Molecular FormulaC9H8O3
InChI KeyQBHDSQZASIBAAI-UHFFFAOYSA-N
SMILESCC(=O)C1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder or crystals
Melting point206°C to 210°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only