Dichloromethane-d2, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
Dichloromethane-d2, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
Thermo Scientific™

Dichloromethane-d2, for NMR, 99.5 atom % D, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 176110500
Price (USD)/ Each
730.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
859.00 
Save 128.35 (15%)
-
Add to cart
Quantity:
50 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)/ Each
730.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
859.00 
Save 128.35 (15%)
Add to cart
Dichloromethane-d2, for NMR, 99.5 atom % D, Thermo Scientific Chemicals
Catalog number176110500
Price (USD)/ Each
730.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
859.00 
Save 128.35 (15%)
-
Add to cart
Chemical Identifiers
CAS1665-00-5
IUPAC Namedichloro(²H₂)methane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-DICFDUPASA-N
SMILES[2H]C([2H])(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.04 % (Coulometric)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
D-Enrichment>=99.4 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only