Thermo Scientific Chemicals

1,4-Dioxa-8-azaspiro[4.5]decane, 98%, Thermo Scientific Chemicals

Catalog number: 176370500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

1,4-Dioxa-8-azaspiro[4.5]decane, 98%, Thermo Scientific Chemicals

Catalog number: 176370500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 176370500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
177-11-7
IUPAC Name
1,4-dioxa-8-azaspiro[4.5]decan-8-ium
Molecular Formula
C7H14NO2
InChI Key
KPKNTUUIEVXMOH-UHFFFAOYSA-O
SMILES
C1COC2(CC[NH2+]CC2)O1
Appearance (Form)
Liquid
Titration with HClO4
>=97.5 %
GC
>=97.5 %
Infrared spectrum
Conforms
Refractive index
1.4809 to 1.4829 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text