4-(Trifluoromethyl)phenol, 98%, Thermo Scientific Chemicals
4-(Trifluoromethyl)phenol, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(Trifluoromethyl)phenol, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Packaging:
Glass bottle
Catalog number 176440250
Price (USD)/ Each
266.65
Online exclusive
314.00 
Save 47.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
266.65
Online exclusive
314.00 
Save 47.35 (15%)
Add to cart
4-(Trifluoromethyl)phenol, 98%, Thermo Scientific Chemicals
Catalog number176440250
Price (USD)/ Each
266.65
Online exclusive
314.00 
Save 47.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name4-(trifluoromethyl)phenol
Molecular FormulaC7H5F3O
InChI KeyBAYGVMXZJBFEMB-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C(F)(F)F
Molecular Weight (g/mol)162.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Infrared spectrumConforms
Appearance (Color)White to yellow-brown
GC>=97.5 %
Melting point40°C to 46°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only