3-Methyl-1,2-cyclopentanedione, 99%, Thermo Scientific Chemicals
3-Methyl-1,2-cyclopentanedione, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Methyl-1,2-cyclopentanedione, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 176460500
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS765-70-8
IUPAC Name3-methylcyclopentane-1,2-dione
Molecular FormulaC6H8O2
InChI KeyOACYKCIZDVVNJL-UHFFFAOYNA-N
SMILESCC1CCC(=O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point104°C to 108°C
GC>=98.5 %
Appearance (Form)Powder or crystals
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only