Thermo Scientific Chemicals

Iodobenzene diacetate, 98%, Thermo Scientific Chemicals

Catalog number: 176565000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Iodobenzene diacetate, 98%, Thermo Scientific Chemicals

Catalog number: 176565000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 176565000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3240-34-4
IUPAC Name
(acetyloxy)(phenyl)-λ³-iodanyl acetate
Molecular Formula
C10H11IO4
InChI Key
ZBIKORITPGTTGI-UHFFFAOYSA-N
SMILES
CC(=O)O[I](OC(C)=O)C1=CC=CC=C1
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration Iodometric
>=97.5 %
Melting point
159°C to 164°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text