4-Formylcinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
4-Formylcinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Formylcinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 176590010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS23359-08-2
IUPAC Name(2E)-3-(4-formylphenyl)prop-2-enoic acid
Molecular FormulaC10H8O3
InChI KeyLBOUHDMYVURTMA-AATRIKPKSA-N
SMILESOC(=O)C=CC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to beige
Titration with NaOH>=97.5 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point250°C to 255°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only