Thermo Scientific Chemicals

4-Formylcinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 176590250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Formylcinnamic acid, 98%, predominantly trans, Thermo Scientific Chemicals

Catalog number: 176590250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 176590250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23359-08-2
IUPAC Name
(2E)-3-(4-formylphenyl)prop-2-enoic acid
Molecular Formula
C10H8O3
InChI Key
LBOUHDMYVURTMA-AATRIKPKSA-N
SMILES
OC(=O)C=CC1=CC=C(C=O)C=C1
Appearance (Color)
Light yellow to beige
Titration with NaOH
>=97.5 %
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
250°C to 255°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text