Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water, Thermo Scientific Chemicals

Catalog number: 176610500
50 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 40 wt.% (1.5M) solution in water, Thermo Scientific Chemicals

Catalog number: 176610500
50 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 176610500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2052-49-5, 7732-18-5
Appearance (Form)
Clear solution
Color scale
=<150 APHA
Titration with HCl
38 to 42 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Phase transfer catalyst
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text