Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT
Thermo Scientific Chemicals

Tetrahydrofuran, 99+%, extra pure, stabilized with BHT

CAS: 109-99-9 | C4H8O | 72.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
25 L
Packaging:
View all
Plastic Drum
Catalog number 176630100
Price (USD)
386.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
10 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
386.00
Each
Add to cart
Chemical Identifiers
CAS109-99-9
IUPAC Nameoxolane
Molecular FormulaC4H8O
InChI KeyWYURNTSHIVDZCO-UHFFFAOYSA-N
SMILESC1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Water=<0.05 % (Coulometric)
Peroxides=<0.0025 % (as H2O2)
Residue after evaporation=<0.005 %
Color scale=<10 APHA
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2544614Certificate of AnalysisJul 14, 2025176630025, 176630010, 176630100, 176630050, 176630250
2543924Certificate of AnalysisJul 08, 2025176630025, 176630010, 176630100, 176630050, 176630250
2536669Certificate of AnalysisMay 24, 2025176630025, 176630010, 176630100, 176630050, 176630250
A0457418Certificate of AnalysisMay 17, 2025176630025, 176630010, 176630100, 176630050, 176630250
2403865Certificate of AnalysisJan 31, 2025176630025, 176630010, 176630100, 176630050, 176630250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection