Thermo Scientific Chemicals

Tetrahydrofuran, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals

Catalog number: 176630250
25 L, Metal Drum, Each
Thermo Scientific Chemicals

Tetrahydrofuran, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals

Catalog number: 176630250
25 L, Metal Drum, Each
Quantity
Packaging
Catalog number: 176630250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
109-99-9
IUPAC Name
oxolane
Molecular Formula
C4H8O
InChI Key
WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES
C1CCOC1
GC
>=99.0 %
Water
=<0.05 % (Coulometric)
Peroxides
=<0.0025 % (as H2O2)
Residue after evaporation
=<0.005 %
Color scale
=<10 APHA

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text