Thermo Scientific Chemicals

Hexanes, 98+%, extra pure, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 176660010
1 L, Glass Bottle, Each
Thermo Scientific Chemicals

Hexanes, 98+%, extra pure, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 176660010
1 L, Glass Bottle, Each
Quantity
Packaging
Catalog number: 176660010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92112-69-1, 110-54-3
Color scale
=<10 APHA
Infrared spectrum
Conforms
GC
>=98.0 % (sum of isomers)
Refractive index
1.3780 to 1.3820 (20°C, 589 nm)
Free acid (CH3COOH)
=<0.002 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text