Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 176688000
Price (USD)/ Each
358.65
Online exclusive
398.00 
Save 39.35 (10%)
-
Add to cart
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)/ Each
358.65
Online exclusive
398.00 
Save 39.35 (10%)
Add to cart
Boron trichloride, 1M solution in methylene chloride, AcroSeal™, Thermo Scientific Chemicals
Catalog number176688000
Price (USD)/ Each
358.65
Online exclusive
398.00 
Save 39.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10294-34-5, 75-09-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to brown
Appearance (Form)Solution
AppearanceMay darken on storage
Titration Argentometric0.9 to 1.1 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only