Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol, 99.9%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
500 mL
1 L
2.5 L
25 L
Catalog number 176840100
Price (USD)/ Each
242.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
285.00 
Save 42.35 (15%)
-
Add to cart
Quantity:
10 L
Request bulk or custom format
Price (USD)/ Each
242.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
285.00 
Save 42.35 (15%)
Add to cart
Methanol, 99.9%, for analysis, Thermo Scientific Chemicals
Catalog number176840100
Price (USD)/ Each
242.65
Special Offer
Online exclusive
Ends: 30-Jun-2025
285.00 
Save 42.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
Molecular Weight (g/mol)32.04
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<0.05 ppm
Potassium (K)=<0.5 ppm
Sodium (Na)=<0.5 ppm
Total phosphorus (P)=<0.05 ppm
Total silicon (Si)=<0.05 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only