Methanol, 99.9%, for analysis
Methanol, 99.9%, for analysis
Methanol, 99.9%, for analysis
Methanol, 99.9%, for analysis
Thermo Scientific Chemicals

Methanol, 99.9%, for analysis

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
500 mL
1 L
2.5 L
10 L
Catalog number 176840250
Price (USD)
391.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
25 L
Request bulk or custom format
Price (USD)
391.00
Each
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.05 % (K.F.)
Carbonyl compounds=<0.001 %
Residue after evaporation=<0.001 %
KMnO4-reducing substance=<0.00025 %
Calcium (Ca)=<0.2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link