Thermo Scientific Chemicals

exo-2-Aminonorbornane, 99%, Thermo Scientific Chemicals

Catalog number: 176950050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

exo-2-Aminonorbornane, 99%, Thermo Scientific Chemicals

Catalog number: 176950050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 176950050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7242-92-4
IUPAC Name
(1R,2R,4S)-bicyclo[2.2.1]heptan-2-amine
Molecular Formula
C7H13N
InChI Key
JEPPYVOSGKWVSJ-YUZWJPFSNA-N
SMILES
N[C@@H]1C[C@H]2CC[C@@H]1C2
Appearance (Color)
Clear colorless to light yellow
Infrared spectrum
Conforms
GC
>=98.5 %
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text