Silver carbonate, 99%, pure
Silver carbonate, 99%, pure
Silver carbonate, 99%, pure
Silver carbonate, 99%, pure
Thermo Scientific Chemicals

Silver carbonate, 99%, pure

CAS: 534-16-7 | CAg2O3 | 275.75 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 176971000
Price (USD)
326.65
Special offer
Online exclusive
384.00
Save 57.35 (15%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
326.65
Special offer
Online exclusive
384.00
Save 57.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS534-16-7
IUPAC Namecarbonic acid silver
Molecular FormulaCH2AgO3
InChI KeyLKZMBDSASOBTPN-UHFFFAOYSA-N
SMILES[Ag].OC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granular powder
Appearance (Color)Yellow to green
Assay>=98.5 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0463132Certificate of AnalysisMay 22, 2025176971000, 176970250
A0468343Certificate of AnalysisJan 31, 2025176971000, 176970250
A0463132Certificate of AnalysisMay 23, 2024176971000, 176970250
A0447986Certificate of AnalysisDec 08, 2023176971000, 176970250
A0442141Certificate of AnalysisAug 25, 2023176971000, 176970250
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection