Thermo Scientific Chemicals

Triethylborane, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 176988000
800 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Triethylborane, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 176988000
800 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 176988000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
97-94-9
IUPAC Name
triethylborane
Molecular Formula
C6H15B
InChI Key
LALRXNPLTWZJIJ-UHFFFAOYSA-N
SMILES
CCB(CC)CC
Appearance (Color)
Colorless to amber
Strength
0.95 to 1.30 M
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text