Potassium hydrogen phthalate, 99.99%, acidimetric standard
Potassium hydrogen phthalate, 99.99%, acidimetric standard
Potassium hydrogen phthalate, 99.99%, acidimetric standard
Potassium hydrogen phthalate, 99.99%, acidimetric standard
Thermo Scientific Chemicals

Potassium hydrogen phthalate, 99.99%, acidimetric standard

CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 177121000
Price (EUR)
39,87
Online Exclusive
46,90
Save 7,03 (15%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
39,87
Online Exclusive
46,90
Save 7,03 (15%)
Each
Add to cart
Chemical Identifiers
CAS877-24-7
IUPAC Namepotassium 2-carboxybenzoate
Molecular FormulaC8H5KO4
InChI KeyIWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES[K+].OC(=O)C1=CC=CC=C1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay99.95 to 100.05 %
pH3.8 to 4 (5 % at 20°C)
Chlorine compounds=<30 ppm (as Cl)
Iron (Fe)=<5 ppm
Sulfate (SO4)=<60 ppm
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link