Thermo Scientific Chemicals

Potassium hydrogen phthalate, 99.99%, acidimetric standard, Thermo Scientific Chemicals

Catalog number: 177125000
500 g, Plastic Bottle, Each
Thermo Scientific Chemicals

Potassium hydrogen phthalate, 99.99%, acidimetric standard, Thermo Scientific Chemicals

Catalog number: 177125000
500 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 177125000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
877-24-7
IUPAC Name
potassium 2-carboxybenzoate
Molecular Formula
C8H5KO4
InChI Key
IWZKICVEHNUQTL-UHFFFAOYSA-M
SMILES
[K+].OC(=O)C1=CC=CC=C1C([O-])=O
Assay
99.95 to 100.05 %
pH
3.8 to 4 (5 % at 20°C)
Chlorine compounds
=<30 ppm (as Cl)
Iron (Fe)
=<5 ppm
Sulfate (SO4)
=<60 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text