Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methanol, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
5 L
Packaging:
View all
Plastic Can
Catalog number 177150250
Price (USD)/ Each
309.65
Online exclusive
364.00 
Save 54.35 (15%)
-
Add to cart
Quantity:
25 L
Packaging:
Plastic Can
Request bulk or custom format
Price (USD)/ Each
309.65
Online exclusive
364.00 
Save 54.35 (15%)
Add to cart
Methanol, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number177150250
Price (USD)/ Each
309.65
Online exclusive
364.00 
Save 54.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
Molecular Weight (g/mol)32.04
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.2 % (K.F.)
Appearance (Form)Clear liquid
GC>=99.0 %
Residue after evaporation=<0.005 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only