Thermo Scientific Chemicals

Tributyl borate, 98%, Thermo Scientific Chemicals

Catalog number: 177191000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

Tributyl borate, 98%, Thermo Scientific Chemicals

Catalog number: 177191000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 177191000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
688-74-4
IUPAC Name
tributyl borate
Molecular Formula
C12H27BO3
InChI Key
LGQXXHMEBUOXRP-UHFFFAOYSA-N
SMILES
CCCCOB(OCCCC)OCCCC
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless
Refractive index
1.4080 to 1.4100 (20°C, 589 nm)
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text