Thermo Scientific Chemicals

3-Bromoaniline, 98%, Thermo Scientific Chemicals

Catalog number: 177211000
100 mL, Glass bottle, Each
Thermo Scientific Chemicals

3-Bromoaniline, 98%, Thermo Scientific Chemicals

Catalog number: 177211000
100 mL, Glass bottle, Each
Quantity
Packaging
Catalog number: 177211000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
591-19-5
IUPAC Name
3-bromoaniline
Molecular Formula
C6H6BrN
InChI Key
DHYHYLGCQVVLOQ-UHFFFAOYSA-N
SMILES
NC1=CC=CC(Br)=C1
Infrared spectrum
Conforms
GC
>=97.5 %
Refractive index
1.6240 to 1.6270 (20°C, 589 nm)
Appearance (Color)
Clear light yellow to orange-brown
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text