Borane-tert-butylamine complex, 95%, powder, Thermo Scientific Chemicals
Borane-tert-butylamine complex, 95%, powder, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Borane-tert-butylamine complex, 95%, powder, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 177300250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7337-45-3
IUPAC Name2-methylpropan-2-amine borane
Molecular FormulaC4H14BN
InChI KeyGKFJEDWZQZKYHV-UHFFFAOYSA-N
SMILESB.CC(C)(C)N
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point96°C to 101°C
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration>=94 % (Active hydrogen)
Appearance (Color)White to off-white
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only